Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYB47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101729.3 | NP_173306.1 | 1 - 2 | 330 | 75 | |
GenBank mRNA | AY065166.1 | - | 1 - 2 | 321 | 75 | |
AY114579.1 | - | 1 - 2 | 134 | 75 | ||
AY519556.1 | - | 1 - 2 | 134 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Arabidopsis thaliana Probe spans exons |