Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAMMA CA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198110.1 | NP_001185039.1 | 1 - 2 | 288 | 95 | |
NM_101815.4 | NP_564091.1 | 1 - 2 | 365 | 95 | ||
GenBank mRNA | AK176887.1 | - | 1 - 2 | 171 | 95 | |
AK229264.1 | - | 1 - 2 | 162 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |