Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HVA22H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101850.2 | NP_564100.1 | 6 - 7 | 753 | 83 | |
GenBank mRNA | AY045614.1 | - | 6 - 7 | 655 | 83 | |
BT001012.1 | - | 5 - 6 | 452 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |