Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YUC10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_103784.2 | NP_175321.1 | 1 - 2 | 613 | 82 | |
GenBank mRNA | DQ056487.1 | - | 1 - 2 | 613 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |