Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATGRIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345737.1 | NP_001332639.1 | 5 - 6 | 1074 | 84 | |
NM_001345738.1 | NP_001332640.1 | 5 - 6 | 1049 | 84 | ||
NM_126000.2 | NP_569025.2 | 4 - 5 | 779 | 84 | ||
NM_180946.3 | NP_851277.1 | 5 - 6 | 1109 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |