Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPK28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085337.1 | NP_001078806.1 | 10 - 11 | 1427 | 98 | |
NM_001345751.1 | NP_001331199.1 | 10 - 11 | 1427 | 98 | ||
NM_126019.4 | NP_201422.1 | 11 - 12 | 1319 | 98 | ||
NM_180949.3 | NP_851280.1 | 10 - 11 | 1427 | 98 | ||
GenBank mRNA | AK227052.1 | - | 10 - 11 | 1252 | 98 | |
AK317109.1 | - | 10 - 11 | 1246 | 98 | ||
AY139991.1 | - | 10 - 11 | 1251 | 98 | ||
BT010380.1 | - | 10 - 11 | 1237 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |