Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPK28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126036.2 | NP_001119508.1 | 10 - 11 | 1714 | 88 | |
NM_126019.4 | NP_201422.1 | 12 - 13 | 1501 | 88 | ||
NM_180949.3 | NP_851280.1 | 11 - 12 | 1609 | 88 | ||
GenBank mRNA | AK227052.1 | - | 11 - 12 | 1434 | 88 | |
AY139991.1 | - | 11 - 12 | 1433 | 88 | ||
BT010380.1 | - | 11 - 12 | 1419 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |