Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPK28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085337.1 | NP_001078806.1 | 8 - 9 | 1261 | 63 | |
NM_001126036.2 | NP_001119508.1 | 8 - 9 | 1261 | 63 | ||
NM_001345751.1 | NP_001331199.1 | 8 - 9 | 1261 | 63 | ||
NM_001345752.1 | NP_001331200.1 | 8 - 9 | 1291 | 63 | ||
NM_126019.4 | NP_201422.1 | 9 - 10 | 1153 | 63 | ||
NM_180949.3 | NP_851280.1 | 8 - 9 | 1261 | 63 | ||
GenBank mRNA | AK227052.1 | - | 8 - 9 | 1086 | 63 | |
AK317109.1 | - | 8 - 9 | 1080 | 63 | ||
AY139991.1 | - | 8 - 9 | 1085 | 63 | ||
BT010380.1 | - | 8 - 9 | 1071 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |