Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOLT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126036.4 | NP_569032.1 | 8 - 9 | 990 | 115 | |
GenBank mRNA | AK227464.1 | - | 8 - 9 | 855 | 115 | |
BT010139.1 | - | 8 - 9 | 723 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |