Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126073.3 | NP_201476.1 | 4 - 5 | 622 | 116 | |
GenBank mRNA | AY099638.1 | - | 4 - 5 | 568 | 116 | |
BT002161.1 | - | 4 - 5 | 469 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |