Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126073.3 | NP_201476.1 | 7 - 8 | 1323 | 87 | |
GenBank mRNA | AY099638.1 | - | 7 - 8 | 1269 | 87 | |
BT002161.1 | - | 7 - 8 | 1170 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |