Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126086.7 | NP_201488.1 | 2 - 3 | 1442 | 68 | |
GenBank mRNA | AB164303.1 | - | 2 - 3 | 1361 | 68 | |
AB473834.1 | - | 2 - 3 | 1217 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Arabidopsis thaliana Probe spans exons |