Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ICU2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345811.1 | NP_001331725.1 | 21 - 22 | 3525 | 115 | |
NM_126110.3 | NP_201511.2 | 21 - 22 | 3507 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |