Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085341.1 | NP_001078810.1 | 2 - 3 | 357 | 72 | |
NM_126111.4 | NP_201512.1 | 2 - 3 | 405 | 72 | ||
GenBank mRNA | BT028946.1 | - | 2 - 3 | 312 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |