Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRAMP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126133.4 | NP_201534.1 | 2 - 3 | 1220 | 70 | |
GenBank mRNA | AY136467.1 | - | 2 - 3 | 757 | 70 | |
BT003406.1 | - | 2 - 3 | 664 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |