Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARLA1D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126156.5 | NP_569051.1 | 5 - 6 | 909 | 136 | |
GenBank mRNA | AY054524.1 | - | 5 - 6 | 661 | 136 | |
AY064616.1 | - | 5 - 6 | 436 | 136 | ||
AY087487.1 | - | 5 - 6 | 627 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |