Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FRO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126159.4 | NP_201560.1 | 2 - 3 | 408 | 87 | |
GenBank mRNA | AK118663.1 | - | 2 - 3 | 299 | 87 | |
BT003675.1 | - | 2 - 3 | 250 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |