Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126173.4 | NP_565252.1 | 1 - 2 | 424 | 101 | |
GenBank mRNA | AB054096.1 | - | 1 - 2 | 254 | 101 | |
AF145045.1 | - | 1 - 2 | 254 | 101 | ||
AF360172.1 | - | 1 - 2 | 274 | 101 | ||
AY056347.1 | - | 1 - 2 | 254 | 101 | ||
AY084784.1 | - | 1 - 2 | 395 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |