Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335061.1 | NP_001323732.1 | 2 - 3 | 1905 | 93 | |
NM_126203.3 | NP_565261.1 | 2 - 3 | 2018 | 93 | ||
NM_179592.2 | NP_849923.1 | 2 - 3 | 2030 | 93 | ||
GenBank mRNA | AF087016.1 | - | 2 - 3 | 1489 | 93 | |
AY091009.1 | - | 2 - 3 | 1589 | 93 | ||
BT008691.1 | - | 2 - 3 | 1594 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |