Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPK17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035862.2 | NP_001030939.1 | 4 - 5 | 680 | 61 | |
NM_001035863.1 | NP_001030940.1 | 4 - 5 | 762 | 61 | ||
NM_001035864.1 | NP_001030941.1 | 4 - 5 | 707 | 61 | ||
NM_126206.4 | NP_178254.2 | 4 - 5 | 782 | 61 | ||
GenBank mRNA | AK228266.1 | - | 4 - 5 | 673 | 61 | |
AK317182.1 | - | 4 - 5 | 680 | 61 | ||
BT006469.1 | - | 3 - 4 | 596 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |