Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPSF73-II | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335074.1 | NP_001325256.1 | 2 - 3 | 526 | 79 | |
NM_126234.3 | NP_178282.2 | 3 - 4 | 440 | 79 | ||
GenBank mRNA | AK221561.1 | - | 3 - 4 | 372 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |