Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATMAP65-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035868.2 | NP_001030945.1 | 6 - 7 | 1069 | 109 | |
NM_126252.4 | NP_178300.3 | 6 - 7 | 2256 | 109 | ||
GenBank mRNA | AK317525.1 | - | 6 - 7 | 1367 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |