Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PP2-B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126284.3 | NP_178332.2 | 2 - 3 | 542 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |