Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLPB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335131.1 | NP_001318183.1 | 3 - 4 | 1262 | 76 | |
NM_001335132.1 | NP_001324185.1 | 3 - 4 | 1262 | 76 | ||
NM_126326.5 | NP_565288.1 | 3 - 4 | 1302 | 76 | ||
NM_179597.1 | NP_849928.1 | 3 - 4 | 1251 | 76 | ||
GenBank mRNA | AB038798.1 | - | 3 - 4 | 1252 | 76 | |
AY045642.1 | - | 3 - 4 | 1054 | 76 | ||
AY058232.1 | - | 3 - 4 | 846 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |