Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126341.3 | NP_178389.1 | 3 - 4 | 754 | 131 | |
GenBank mRNA | AJ289165.1 | - | 3 - 4 | 431 | 131 | |
AK226970.1 | - | 3 - 4 | 558 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |