Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126363.7 | NP_565294.1 | 2 - 3 | 333 | 66 | |
GenBank mRNA | AY065169.1 | - | 2 - 3 | 295 | 66 | |
AY114571.1 | - | 2 - 3 | 162 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |