Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111003.5 | NP_186786.2 | 3 - 4 | 1353 | 108 | |
GenBank mRNA | AY136347.1 | - | 3 - 4 | 1353 | 108 | |
BT010594.1 | - | 3 - 4 | 1331 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |