Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MBD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111012.4 | NP_186795.1 | 6 - 7 | 3760 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |