Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BET10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337357.1 | NP_001326912.1 | 2 - 3 | 1372 | 95 | |
NM_111043.3 | NP_566151.1 | 2 - 3 | 1383 | 95 | ||
GenBank mRNA | AY057662.1 | - | 2 - 3 | 1321 | 95 | |
AY099729.1 | - | 2 - 3 | 1328 | 95 | ||
AY128906.1 | - | 1 - 2 | 1073 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |