Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KUP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111071.3 | NP_186854.1 | 4 - 5 | 789 | 66 | |
GenBank mRNA | AY062747.1 | - | 4 - 5 | 786 | 66 | |
BT003375.1 | - | 4 - 5 | 587 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |