Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LBD41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111122.5 | NP_566175.1 | 1 - 2 | 669 | 61 | |
GenBank mRNA | AB473871.1 | - | 1 - 2 | 508 | 61 | |
AF447895.1 | - | 1 - 2 | 406 | 61 | ||
AY090370.1 | - | 1 - 2 | 380 | 61 | ||
AY122896.1 | - | 1 - 2 | 193 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |