Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VTC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035549.1 | NP_001030626.1 | 10 - 11 | 820 | 76 | |
NM_001125086.1 | NP_001118558.1 | 10 - 11 | 817 | 76 | ||
NM_111155.3 | NP_186936.1 | 10 - 11 | 906 | 76 | ||
GenBank mRNA | AY035150.1 | - | 10 - 11 | 751 | 76 | |
AY063021.1 | - | 10 - 11 | 663 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |