Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084632.1 | NP_001078101.1 | 17 - 18 | 3106 | 97 | |
NM_001202869.2 | NP_001189798.1 | 20 - 21 | 3469 | 97 | ||
NM_001337464.1 | NP_001327050.1 | 14 - 15 | 2587 | 97 | ||
NM_111200.5 | NP_566199.4 | 21 - 22 | 3711 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |