Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341360.1 | NP_001327978.1 | 2 - 3 | 337 | 104 | |
NM_001341361.1 | NP_001327977.1 | 2 - 3 | 414 | 104 | ||
NM_118084.5 | NP_193699.1 | 2 - 3 | 414 | 104 | ||
GenBank mRNA | AB038491.1 | - | 2 - 3 | 115 | 104 | |
AF370309.1 | - | 2 - 3 | 241 | 104 | ||
AY052670.1 | - | 2 - 3 | 277 | 104 | ||
AY063095.1 | - | 2 - 3 | 115 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |