Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AK-HSDH II | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118091.3 | NP_193706.2 | 16 - 17 | 2591 | 83 | |
NM_202847.3 | NP_974576.1 | 16 - 17 | 2632 | 83 | ||
GenBank mRNA | AK228252.1 | - | 16 - 17 | 2560 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |