Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HEN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203853.2 | NP_001190782.1 | 8 - 9 | 2366 | 80 | |
NM_118209.3 | NP_567616.1 | 7 - 8 | 2511 | 80 | ||
GenBank mRNA | AF327068.1 | - | 7 - 8 | 2293 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |