Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HEN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203853.2 | NP_001190782.1 | 9 - 10 | 2481 | 66 | |
NM_118209.3 | NP_567616.1 | 8 - 9 | 2626 | 66 | ||
GenBank mRNA | AF327068.1 | - | 8 - 9 | 2408 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |