Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRCA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118225.5 | NP_193839.4 | 5 - 6 | 934 | 71 | |
GenBank mRNA | AF515728.1 | - | 5 - 6 | 801 | 71 | |
AY081328.1 | - | 5 - 6 | 847 | 71 | ||
BT010547.1 | - | 5 - 6 | 751 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |