Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATERDJ2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341475.1 | NP_001328817.1 | 8 - 9 | 1111 | 106 | |
NM_118237.4 | NP_567621.1 | 8 - 9 | 1270 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |