Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341491.1 | NP_001329575.1 | 7 - 8 | 1591 | 94 | |
NM_118269.6 | NP_193880.5 | 7 - 8 | 1517 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |