Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118281.4 | NP_193892.1 | 3 - 4 | 475 | 71 | |
GenBank mRNA | BT011760.1 | - | 3 - 4 | 475 | 71 | |
BT012384.1 | - | 3 - 4 | 382 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |