Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATML1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036615.3 | NP_001031692.1 | 10 - 11 | 2915 | 58 | |
NM_001341510.1 | NP_001329394.1 | 10 - 11 | 3574 | 58 | ||
NM_001341511.1 | NP_001329395.1 | 11 - 12 | 2925 | 58 | ||
NM_118295.7 | NP_193906.2 | 10 - 11 | 2667 | 58 | ||
GenBank mRNA | AK229970.1 | - | 10 - 11 | 2768 | 58 | |
AY091104.1 | - | 10 - 11 | 2559 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |