Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPK15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203865.1 | NP_001190794.1 | 5 - 6 | 1392 | 91 | |
NM_001341523.1 | NP_001329845.1 | 5 - 6 | 1459 | 91 | ||
NM_118315.2 | NP_193925.1 | 5 - 6 | 1459 | 91 | ||
GenBank mRNA | AY034995.1 | - | 5 - 6 | 1387 | 91 | |
AY063004.1 | - | 5 - 6 | 1347 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |