Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATCES1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118359.4 | NP_567660.1 | 2 - 3 | 545 | 113 | |
GenBank mRNA | AB063253.1 | - | 2 - 3 | 488 | 113 | |
BT008549.1 | - | 1 - 2 | 254 | 113 | ||
BT008652.1 | - | 2 - 3 | 447 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |