Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LDOX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036623.1 | NP_001031700.1 | 2 - 3 | 536 | 103 | |
NM_001341563.1 | NP_001320035.1 | 1 - 2 | 647 | 103 | ||
NM_118417.2 | NP_194019.1 | 2 - 3 | 546 | 103 | ||
GenBank mRNA | AK226417.1 | - | 2 - 3 | 547 | 103 | |
AK317414.1 | - | 2 - 3 | 536 | 103 | ||
AY088203.1 | - | 2 - 3 | 546 | 103 | ||
U70478.1 | - | 1 - 2 | 528 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |