Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ESP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203875.2 | NP_001190804.1 | 13 - 14 | 3121 | 101 | |
NM_001341570.1 | NP_001328112.1 | 13 - 14 | 2968 | 101 | ||
NM_118426.4 | NP_194028.2 | 13 - 14 | 3063 | 101 | ||
GenBank mRNA | AY823256.1 | - | 13 - 14 | 2965 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |