Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ESP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341570.1 | NP_001328112.1 | 22 - 23 | 5008 | 74 | |
NM_118426.4 | NP_194028.2 | 22 - 23 | 5103 | 74 | ||
GenBank mRNA | AY823256.1 | - | 22 - 23 | 5005 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |