Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UTR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084964.1 | NP_001078433.1 | 5 - 6 | 1020 | 105 | |
NM_118430.5 | NP_194032.2 | 5 - 6 | 1049 | 105 | ||
GenBank mRNA | AK227353.1 | - | 5 - 6 | 882 | 105 | |
BT024728.1 | - | 5 - 6 | 698 | 105 | ||
DQ015901.1 | - | 5 - 6 | 749 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |