Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036624.1 | NP_001031701.1 | 7 - 8 | 1024 | 83 | |
NM_001203879.2 | NP_001190808.1 | 7 - 8 | 1420 | 83 | ||
NM_118439.4 | NP_194041.1 | 7 - 8 | 1290 | 83 | ||
GenBank mRNA | AF419576.1 | - | 7 - 8 | 1006 | 83 | |
AY143970.1 | - | 6 - 7 | 835 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |