Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDPK6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118496.4 | NP_194096.1 | 7 - 8 | 1643 | 128 | |
GenBank mRNA | AF385710.1 | - | 7 - 8 | 1451 | 128 | |
AY081732.1 | - | 7 - 8 | 1407 | 128 | ||
AY087637.1 | - | 7 - 8 | 1447 | 128 | ||
BT000736.1 | - | 7 - 8 | 1409 | 128 | ||
U20623.1 | - | 7 - 8 | 1422 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |